missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, Certified, 50% (w/v) ±1%, LabChem™

Supplier:  LabChem LC241405

 Afficher plus de versions de ce produit

Numéro de catalogue. LC241405

Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Spécifications

Spécifications

Sodium Hydroxide
1310-73-2,7732-18-5
HNaO
UN1824
Soluble in water
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
40
Certified
Poly Drum
Liquid
Colorless
64.03,35.97
NaOH
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
20 L
[OH-].[Na+]
39.997
CHEBI:32145
50% ±1% (w/v)
Passes Test
Traceable to NIST

Identifiants chimiques

1310-73-2
39.997
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

Recommended Storage : Room Temperature

FDS
Documents

Documents

Promotions

Promotions