missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 6.00 N, Ricca Chemical
$110.97 - $400.80
Chemical Identifiers
| CAS | 1310-73-2 |
|---|---|
| Molecular Formula | HNaO |
| Molecular Weight (g/mol) | 39.997 |
| InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
| Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
| PubChem CID | 14798 |
| ChEBI | CHEBI:32145 |
| IUPAC Name | sodium;hydroxide |
| SMILES | [OH-].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
746616
|
Ricca Chemical Company
746616 |
500 mL | Natural Poly Bottle |
Each for $110.97
|
|
||||
|
7466-32
|
Ricca Chemical Company
746632 |
1 L | Natural Poly Bottle |
Each for $148.33
|
|
||||
|
74661
|
Ricca Chemical Company
74661 |
4 L | Natural Poly Bottle |
Each for $339.30
|
|
||||
|
74665
|
Ricca Chemical Company
74665 |
20 L | Cubitainer |
Each for $400.80
|
|
||||
Chemical Identifiers
| 1310-73-2 | |
| 39.997 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| CHEBI:32145 | |
| [OH-].[Na+] |
| HNaO | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| 14798 | |
| sodium;hydroxide |
Specifications
| Colorless | |
| 66.0,31.88 | |
| HNaO | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| sodium;hydroxide | |
| 14798 | |
| Laboratory | |
| >13 | |
| Liquid | |
| Sodium Hydroxide |