missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 6.00 N, Ricca Chemical
Supplier: Ricca Chemical Company 74661
Specifications
| Sodium Hydroxide | |
| 1310-73-2 , 7732-18-5 | |
| 69.0,33.18 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| [OH-].[Na+] | |
| 39.997 | |
| CHEBI:32145 | |
| Natural Poly Bottle | |
| 4 L | |
| 6.00N |
| Colorless | |
| 66.0,31.88 | |
| HNaO | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| sodium;hydroxide | |
| 14798 | |
| Laboratory | |
| >13 | |
| Liquid |
Chemical Identifiers
| 1310-73-2 | |
| 39.997 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| CHEBI:32145 | |
| [OH-].[Na+] |
| HNaO | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| 14798 | |
| sodium;hydroxide |