missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 6.00 N, Ricca Chemical

Supplier:  Ricca Chemical Company 74665

 View more versions of this product

Catalog No. 74665


Only null left

Description

Description

Specifications

Specifications

Sodium Hydroxide
1310-73-2 , 7732-18-5
69.0,33.18
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
[OH-].[Na+]
39.997
CHEBI:32145
Cubitainer
20 L
6.00N
Colorless
66.0,31.88
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
Laboratory
>13
Liquid

Chemical Identifiers

1310-73-2
39.997
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions