missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 50% (w/w) Solution, Technical, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S62220LT

 Afficher plus de versions de ce produit

Numéro de catalogue. 18613625


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Spécifications

Spécifications

Sodium Hydroxide Solution
50%
Chemical Solution
1310-73-2,7732-18-5
20 L
Poly Drum

Identifiants chimiques

1310-73-2
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.