missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 0.5 N Aqueous Solution, Spectrum™ Chemical

S-360, 1310-73-2

$152.73 - $834.50

Identifiants chimiques

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 40.00
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
IUPAC Name sodium hydroxide
SMILES [OH-].[Na+]
voir les spécifications

Produits 4
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
18606535
Afficher les documents
Spectrum Chemical Mfg Cor
S360500ML
500 mL
chaque for $152.73
Il en reste null
 
18606532
Afficher les documents
Spectrum Chemical Mfg Cor
S3601LT
1 L
chaque for $191.73
Il en reste null
 
18606533
Afficher les documents
Spectrum Chemical Mfg Cor
S3604LTPL
4 L
chaque for $347.83
Il en reste null
 
18606534
Afficher les documents
Spectrum Chemical Mfg Cor
S36020LT
20 L
chaque for $834.50
Il en reste null
 
Description

Description

Spectrum™ Chemical Sodium Hydroxide, 0.5 N Aqueous Solution - Spectrum solutions utilize the highest quality raw materials appropriate to your product. Most raw materials meet or exceed the specifications established by the American Chemical Society. Where appropriate, many finished products are traceable to NIST Standard Reference Materials. Manufacturing, quality control testing, and packaging are all performed in Spectrum’s own facilities. Record keeping and sample retention of all produced lots ensure product consistency and complete traceability.

Spécifications

Identifiants chimiques

1310-73-2
40.00
sodium hydroxide
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
[OH-].[Na+]

Spécifications

1310-73-2,7732-18-5
HNaO
500 mL
[OH-].[Na+]
40.00
Plastic
0.02,0.98
0.5 N ± 0.005 N
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide
Chemical Solution
Sodium Hydroxide Solution
Videos
FDS
missing translation for 'documents'

missing translation for 'documents'