missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hyaluronate, 95%
CAS: 9067-32-7 | (C14H20NO11Na)n | 417.30 g/mol
Supplier: Thermo Scientific Chemicals 251770050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
| Sodium hyaluronate | |
| White to Cream | |
| 10% max. (500mg, 105°C, 6 hrs) | |
| 94% min. (On dry substance) | |
| (C14H20NO11Na)n | |
| MFCD00875848 | |
| 15,4796 | |
| Solubility in water: soluble. | |
| CC(=O)NC1C(OC2C(O)C(O)C(O-*)OC2C(=O)O[Na])OC(CO)C(O)C1O-* | |
| 417.30 | |
| Powder |
| 9067-32-7 | |
| 6.0 to 8.0 (0.5% aq. soln.) | |
| Authentic | |
| Glass bottle | |
| (C14H20NO11Na)n | |
| 5 g | |
| hyaluronate tetrasaccharide, d0e9sz, 6-3-acetamido-2-6-3-acetamido-2,5-dihydroxy-6-hydroxymethyl oxan-4-yl oxy-2-carboxy-4,5-dihydroxyoxan-3-yl oxy-5-hydroxy-6-hydroxymethyl oxan-4-yl oxy-3,4,5-trihydroxyoxane-2-carboxylic acid | |
| YWIVKILSMZOHHF-QJZPQSOGSA-N | |
| Sodium hyaluronate | |
| 95% |
Identifiants chimiques
| 9067-32-7 | |
| 417.30 | |
| YWIVKILSMZOHHF-QJZPQSOGSA-N | |
| Sodium hyaluronate |
| (C14H20NO11Na)n | |
| MFCD00875848 | |
| hyaluronate tetrasaccharide, d0e9sz, 6-3-acetamido-2-6-3-acetamido-2,5-dihydroxy-6-hydroxymethyl oxan-4-yl oxy-2-carboxy-4,5-dihydroxyoxan-3-yl oxy-5-hydroxy-6-hydroxymethyl oxan-4-yl oxy-3,4,5-trihydroxyoxane-2-carboxylic acid | |
| CC(=O)NC1C(OC2C(O)C(O)C(O-*)OC2C(=O)O[Na])OC(CO)C(O)C1O-* |
RUO – Research Use Only