missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexametaphosphate, tech.

Numéro de catalogue. AA14706A7
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AA14706A7 5 kg
AA14706A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AA14706A7 Fournisseur Thermo Scientific Chemicals Code fournisseur 014706A7
Il en reste null

CAS: 10124-56-8 | Na6O18P6

Sodium hexametaphosphate is a food additive used in dairy product, canned milk, packaged egg whites, ice cream, seafood and meat processing. It is also used as a sequestrant and water softening agent and detergents. It is an active ingredient in toothpastes as an anti-staining agent. It prevents the steel corrosion. Further, it is used in building, water treatment and in photographic developing. In addition, it is used as a retarder in dental alginate impression materials and as a thinner for petroleum drilling fluids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 10124-56-8
Molecular Formula Na6O18P6
MDL Number MFCD00136045
Synonym Sodium metaphosphate

Spécifications

Boiling Point 1500°C
Melting Point 628°C
Color White
Physical Form Granules
Quantity 5 kg
Solubility Information Soluble in water. Insoluble in organic solvents
Formula Weight 611.77
Grade Technical
Odor Odorless
Refractive Index 1.482
Appearance White granules
Chemical Name or Material Sodium hexametaphosphate
Afficher plus de résultats Afficher moins de résultats
Hazard Category H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
EINECSNumber 233-343-1
RTECSNumber OY3675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only