missing translation for 'onlineSavingsMsg'
Learn More

Sodium bicarbonate, Honeywell Fluka™

Numéro de catalogue. 6003175
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
6 x 500 g
12 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
6003175 6 x 500 g
6003172 500 g
6003168 1 kg
6003169 2.5 kg
6003167 12 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Numéro de catalogue. 6003175 Fournisseur Honeywell Chemicals Code fournisseur S60146X500G
Il en reste null

ACS Reagent,≥99.7%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Identifiants chimiques

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
PubChem CID 516892
ChEBI CHEBI:32139
SMILES [Na+].OC([O-])=O

Spécifications

Quantity 6 x 500 g
Linear Formula NaHCO3
UN Number NONH for all modes of transport
Beilstein 4153970
IUPAC Name sodium;hydrogen carbonate
Formula Weight 84.01g/mol
Percent Purity 99.7 to 100.3%
Chemical Name or Material Sodium bicarbonate
RTECSNumber VZ0950000