missing translation for 'onlineSavingsMsg'
Learn More

Sodium Aluminate, Technical, Spectrum™ Chemical

Numéro de catalogue. p-7158614
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
18606291 500 g Poly Bottle
18606292 2.5 kg Poly Bottle
18606293 12 kg Poly Pail
18606294 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. 18606291 Fournisseur Spectrum Chemical Mfg Cor Code fournisseur S1120500GM
Il en reste null

S1120, 1302-42-7, Na2O·Al2O3·3H2O

Spectrum™ Chemical Sodium Aluminate, Technical is an inorganic chemical that is used as in water softening systems, as a coagulant aid to improve flocculation, and for removing dissolved silica and phosphates. Technical grade products supplied by Spectrum are indicative of a grade suitable for general industrial use. These materials may or may not have a Certificate of Analysis available.

Identifiants chimiques

CAS 1302-42-7
Molecular Formula AlNaO2
Molecular Weight (g/mol) 81.97
InChI Key IYJYQHRNMMNLRH-UHFFFAOYSA-N
IUPAC Name sodium dioxoalumanuide
SMILES [Na+].O=[Al-]=O

Spécifications

CAS Max % 1
Quantity 500 g
UN Number UN2812
Grade Technical
Packaging Poly Bottle