Learn More
Simport™ Scientific T100 BioTube™ Racks
The T100 BioTube™ Racks rack with the standard 96-well on-center spacing of tubes offers a color-coding system using colored interchangeable plastic grids.
Supplier:  Simport™ Scientific T1003B
 Afficher plus de versions de ce produit
Numéro de catalogue. 05-500-8
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Made of polypropylene
  • Free of RNase, DNase, Pyrogen, and DNA
  • Convenient carrying handles on both sides
  • Cover, grid plate, and base are keyed to prevent misalignment
  • Easy to read ID numbers and letters
  • Grid plate stands on 4 legs and can be placed on a lab counter or in a water bath
  • These racks and tubes are also ideal for storing, freezing, and transporting reagents and specimens
Spécifications
Spécifications
Blue
HTLV-III testing, bacterial and hybridoma cell uptake studies, cell harvesting
10/Cs.
12-strip Tube Rack
Polypropylene
Rack with 12 strips of 8 tubes
Non-sterile
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS