Learn More
Simport™ Scientific T100 BioTube™ Racks
The T100 BioTube™ Racks rack with the standard 96-well on-center spacing of tubes offers a color-coding system using colored interchangeable plastic grids.
Supplier: Simport™ Scientific T1001R
Description
- Made of polypropylene
- Free of RNase, DNase, Pyrogen, and DNA
- Convenient carrying handles on both sides
- Cover, grid plate, and base are keyed to prevent misalignment
- Easy to read ID numbers and letters
- Grid plate stands on 4 legs and can be placed on a lab counter or in a water bath
- These racks and tubes are also ideal for storing, freezing, and transporting reagents and specimens
Spécifications
| Red | |
| HTLV-III testing, bacterial and hybridoma cell uptake studies, cell harvesting | |
| 10/Cs. | |
| 96-plain Tube Rack |
| Polypropylene | |
| Rack with 96 plain individual tubes | |
| Non-sterile |