missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate Solution, 1.0M, Honeywell Fluka™

Volumetric, 1.0 M AgNO3 (1.0N)

$564.13 - $1758.20

Identifiants chimiques

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
6001337
Afficher les documents
Honeywell Chemicals
319449500ML
500 mL
chaque for $564.13
Il en reste null
 
6001336
Afficher les documents
Honeywell Chemicals
3194491L
1 L
chaque for $1,758.20
Il en reste null
 
Description

Description

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

Spécifications

Identifiants chimiques

7761-88-8
169.87
SQGYOTSLMSWVJD-UHFFFAOYSA-N
24470
silver(1+) nitrate
AgNO3
MFCD00003414
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
CHEBI:32130
[Ag+].[O-][N+]([O-])=O

Spécifications

7761-88-8
AgNO3
3264
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver(1+) nitrate
24470
169.87g/mol
1.135g/mL at 25°C; Vapor Density: 5.8 (vs air)
500 mL
MFCD00003414
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
[Ag+].[O-][N+]([O-])=O
169.87
CHEBI:32130
1.0M
Silver Nitrate Solution
Videos
Sécurité et manipulation

Sécurité et manipulation

P220-P273-P280-P305 + P351 + P338-P310-P501

H314-H410

FDS
missing translation for 'documents'

missing translation for 'documents'