missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Nitrate, For Chloride, (1mL = 0.5mg Cl-), Certified, 0.0141N ±0.0002N (0.0141M), LabChem™
$98.39 - $1271.07
Chemical Identifiers
| CAS | 7761-88-8 |
|---|---|
| Molecular Formula | AgNO3 |
| Molecular Weight (g/mol) | 169.87 |
| MDL Number | MFCD00003414 |
| InChI Key | SQGYOTSLMSWVJD-UHFFFAOYSA-N |
| Synonym | silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol |
| PubChem CID | 24470 |
| ChEBI | CHEBI:32130 |
| IUPAC Name | silver(1+) nitrate |
| SMILES | [Ag+].[O-][N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
LC226001
|
LabChem
LC226001 |
500 mL |
Each for $98.39
|
|
|||||
|
LC226002
|
LabChem
LC226002 |
1 L |
Each for $165.61
|
|
|||||
|
LC226004
|
LabChem
LC226004 |
4 L |
Each for $328.00
|
|
|||||
|
LC226006
|
LabChem
LC226006 |
10 L |
Each for $681.50
|
|
|||||
|
LC226005
|
LabChem
LC226005 |
20 L |
Each for $1,271.07
|
|
|||||
Chemical Identifiers
| 7761-88-8 | |
| 169.87 | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| 24470 | |
| silver(1+) nitrate |
| AgNO3 | |
| MFCD00003414 | |
| silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
| CHEBI:32130 | |
| [Ag+].[O-][N+]([O-])=O |
Specifications
| Colorless | |
| 99.76,0.24 | |
| AgNO3 | |
| 0.0141N ±0.0002N (0.0141M) | |
| silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| silver(1+) nitrate | |
| 24470 | |
| 169.87 | |
| Passes Test | |
| 1g/mL | |
| Nitrogen oxides; Fume; Carbon monoxide; Carbon dioxide | |
| Liquid |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature