missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate, For Chloride, (1mL = 0.5mg Cl-), Certified, 0.0141N ±0.0002N (0.0141M), LabChem™

Supplier:  LabChem LC226005

 View more versions of this product

Catalog No. LC226005


Only null left

Description

Description

Specifications

Specifications

Silver Nitrate
For Chloride, (1mL = 0.5mg Cl-)
99.76,0.24
AgNO3
0.0141N ±0.0002N (0.0141M)
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver(1+) nitrate
24470
169.87
Passes Test
1g/mL
Nitrogen oxides; Fume; Carbon monoxide; Carbon dioxide
Liquid
Colorless
7761-88-8 , 7732-18-5
AgNO3
MFCD00003414
1g/mL
Soluble in water
[Ag+].[O-][N+]([O-])=O
169.87
CHEBI:32130
Certified
Cubitainer™
Traceable to NIST
20 L

Chemical Identifiers

7761-88-8
169.87
SQGYOTSLMSWVJD-UHFFFAOYSA-N
24470
silver(1+) nitrate
AgNO3
MFCD00003414
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
CHEBI:32130
[Ag+].[O-][N+]([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions