CAS RN 7761-88-8
Silver Nitrate Solution (0.1N (N/10)/Certified), Fisher Chemical™
CAS: 7761-88-8 Molecular Formula: AgNO3 Molecular Weight (g/mol): 169.87 MDL Number: MFCD00003414 InChI Key: SQGYOTSLMSWVJD-UHFFFAOYSA-N Synonym: silver nitrate,silvernitrate,lunar caustic,silbernitrat,argenti nitras,nitrate d'argent,nitric acid silver i salt,silver mononitrate,silver i nitrate,argerol PubChem CID: 24470 ChEBI: CHEBI:32130 IUPAC Name: silver(1+) nitrate SMILES: [Ag+].[O-][N+]([O-])=O
Silver Nitrate (Crystalline/Certified ACS), Fisher Chemical™
CAS: 7761-88-8 Molecular Formula: AgNO3 Molecular Weight (g/mol): 169.87 MDL Number: MFCD00003414 InChI Key: SQGYOTSLMSWVJD-UHFFFAOYSA-N Synonym: silver nitrate,silvernitrate,lunar caustic,silbernitrat,argenti nitras,nitrate d'argent,nitric acid silver i salt,silver mononitrate,silver i nitrate,argerol PubChem CID: 24470 ChEBI: CHEBI:32130 IUPAC Name: silver(1+) nitrate SMILES: [Ag+].[O-][N+]([O-])=O
Silver Nitrate Solution (1N/Certified), Fisher Chemical
CAS: 7761-88-8 Molecular Formula: AgNO3 Molecular Weight (g/mol): 169.87 MDL Number: MFCD00003414 InChI Key: SQGYOTSLMSWVJD-UHFFFAOYSA-N Synonym: silver nitrate,silvernitrate,lunar caustic,silbernitrat,argenti nitras,nitrate d'argent,nitric acid silver i salt,silver mononitrate,silver i nitrate,argerol PubChem CID: 24470 ChEBI: CHEBI:32130 IUPAC Name: silver(1+) nitrate SMILES: [Ag+].[O-][N+]([O-])=O
Silver Nitrate, 0.0100 N (N/100), Ricca Chemical
Molecular Formula: AgNO3 Molecular Weight (g/mol): 169.87 MDL Number: MFCD00003414 InChI Key: SQGYOTSLMSWVJD-UHFFFAOYSA-N Synonym: silver nitrate,silvernitrate,lunar caustic,silbernitrat,argenti nitras,nitrate d'argent,nitric acid silver i salt,silver mononitrate,silver i nitrate,argerol PubChem CID: 24470 ChEBI: CHEBI:32130 IUPAC Name: silver(1+) nitrate SMILES: [Ag+].[O-][N+]([O-])=O
Silver Nitrate, 0.0100 N (N/100) in Isopropyl Alcohol, Ricca Chemical
CAS: 7761-88-8 Molecular Formula: AgNO3 Molecular Weight (g/mol): 169.87 MDL Number: MFCD00003414 InChI Key: SQGYOTSLMSWVJD-UHFFFAOYSA-N Synonym: silver nitrate,silvernitrate,lunar caustic,silbernitrat,argenti nitras,nitrate d'argent,nitric acid silver i salt,silver mononitrate,silver i nitrate,argerol PubChem CID: 24470 ChEBI: CHEBI:32130 IUPAC Name: silver(1+) nitrate SMILES: [Ag+].[O-][N+]([O-])=O
Silver Nitrate, 0.171 N, 1 mL = 10 mg NaCl, Ricca Chemical
CAS: 7761-88-8 Molecular Formula: AgNO3 Molecular Weight (g/mol): 169.87 MDL Number: MFCD00003414 InChI Key: SQGYOTSLMSWVJD-UHFFFAOYSA-N Synonym: silver nitrate,silvernitrate,lunar caustic,silbernitrat,argenti nitras,nitrate d'argent,nitric acid silver i salt,silver mononitrate,silver i nitrate,argerol PubChem CID: 24470 ChEBI: CHEBI:32130 IUPAC Name: silver(1+) nitrate SMILES: [Ag+].[O-][N+]([O-])=O
Silver Nitrate, in Isopropanol, Certified, 0.0100N ±0.0002N (0.01M), LabChem™
CAS: 7761-88-8 Molecular Formula: AgNO3 Molecular Weight (g/mol): 169.87 MDL Number: MFCD00003414 InChI Key: SQGYOTSLMSWVJD-UHFFFAOYSA-N Synonym: silver nitrate,silvernitrate,lunar caustic,silbernitrat,argenti nitras,nitrate d'argent,nitric acid silver i salt,silver mononitrate,silver i nitrate,argerol PubChem CID: 24470 ChEBI: CHEBI:32130 IUPAC Name: silver(1+) nitrate SMILES: [Ag+].[O-][N+]([O-])=O
Silver Nitrate Solution (0.25N), Fisher Chemical™
CAS: 7761-88-8 Molecular Formula: AgNO3 Molecular Weight (g/mol): 169.87 MDL Number: MFCD00003414 InChI Key: SQGYOTSLMSWVJD-UHFFFAOYSA-N Synonym: silver nitrate,silvernitrate,lunar caustic,silbernitrat,argenti nitras,nitrate d'argent,nitric acid silver i salt,silver mononitrate,silver i nitrate,argerol PubChem CID: 24470 ChEBI: CHEBI:32130 IUPAC Name: silver(1+) nitrate SMILES: [Ag+].[O-][N+]([O-])=O
Silver Nitrate (Crystalline/USP), Fisher Chemical™
CAS: 7761-88-8 Molecular Formula: AgNO3 Molecular Weight (g/mol): 169.87 MDL Number: MFCD00003414 InChI Key: SQGYOTSLMSWVJD-UHFFFAOYSA-N Synonym: silver nitrate,silvernitrate,lunar caustic,silbernitrat,argenti nitras,nitrate d'argent,nitric acid silver i salt,silver mononitrate,silver i nitrate,argerol PubChem CID: 24470 ChEBI: CHEBI:32130 IUPAC Name: silver(1+) nitrate SMILES: [Ag+].[O-][N+]([O-])=O
Silver nitrate, ACS, 99.9+% (metals basis)
CAS: 7761-88-8 Molecular Formula: AgNO3 Molecular Weight (g/mol): 169.87 MDL Number: MFCD00003414 InChI Key: SQGYOTSLMSWVJD-UHFFFAOYSA-N Synonym: silver nitrate,silvernitrate,lunar caustic,silbernitrat,argenti nitras,nitrate d'argent,nitric acid silver i salt,silver mononitrate,silver i nitrate,argerol PubChem CID: 24470 ChEBI: CHEBI:32130 IUPAC Name: silver(1+) nitrate SMILES: [Ag+].[O-][N+]([O-])=O
Silver Nitrate, Certified, 0.1000N ±0.0005N (0.1M), LabChem™
CAS: 7761-88-8 Molecular Formula: AgNO3 Molecular Weight (g/mol): 169.87 MDL Number: MFCD00003414 InChI Key: SQGYOTSLMSWVJD-UHFFFAOYSA-N Synonym: silver nitrate,silvernitrate,lunar caustic,silbernitrat,argenti nitras,nitrate d'argent,nitric acid silver i salt,silver mononitrate,silver i nitrate,argerol PubChem CID: 24470 ChEBI: CHEBI:32130 IUPAC Name: silver(1+) nitrate SMILES: [Ag+].[O-][N+]([O-])=O
Invitrogen™ OCT1 (POU2F1) Recombinant Rabbit Monoclonal Antibody (13H18L58)
Rabbit Recombinant Monoclonal Antibody
Metals on Filter Paper, MilliporeSigma™ Supelco™
This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.
TCLP Metals in Soil - PT, MilliporeSigma™ Supelco™
This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.