missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Nitrate, For Chloride, (1mL = 0.5mg Cl-), Certified, 0.0141N ±0.0002N (0.0141M), LabChem™
Supplier: LabChem LC226004
Specifications
| Silver Nitrate | |
| For Chloride, (1mL = 0.5mg Cl-) | |
| 99.76,0.24 | |
| AgNO3 | |
| 0.0141N ±0.0002N (0.0141M) | |
| silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| silver(1+) nitrate | |
| 24470 | |
| 169.87 | |
| Passes Test | |
| 1g/mL | |
| Nitrogen oxides; Fume; Carbon monoxide; Carbon dioxide | |
| Liquid |
| Colorless | |
| 7761-88-8 , 7732-18-5 | |
| AgNO3 | |
| MFCD00003414 | |
| 1g/mL | |
| Soluble in water | |
| [Ag+].[O-][N+]([O-])=O | |
| 169.87 | |
| CHEBI:32130 | |
| Certified | |
| Amber Glass | |
| Traceable to NIST | |
| 4 L |
Chemical Identifiers
| 7761-88-8 | |
| 169.87 | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| 24470 | |
| silver(1+) nitrate |
| AgNO3 | |
| MFCD00003414 | |
| silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
| CHEBI:32130 | |
| [Ag+].[O-][N+]([O-])=O |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature