missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Nitrate, 0.171 N, 1 mL = 10 mg NaCl, Ricca Chemical
For Chloride Titrations
$361.28 - $7489.64
Chemical Identifiers
| CAS | 7761-88-8 |
|---|---|
| Molecular Formula | AgNO3 |
| Molecular Weight (g/mol) | 169.87 |
| MDL Number | MFCD00003414 |
| InChI Key | SQGYOTSLMSWVJD-UHFFFAOYSA-N |
| Synonym | silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol |
| PubChem CID | 24470 |
| ChEBI | CHEBI:32130 |
| IUPAC Name | silver(1+) nitrate |
| SMILES | [Ag+].[O-][N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
700316
|
Ricca Chemical Company
700316 |
500 mL | Amber Poly Bottle |
Each for $361.28
|
|
||||
|
7003-32
|
Ricca Chemical Company
700332 |
1 L | Amber Poly Bottle |
Each for $599.06
|
|
||||
|
7003-1
|
Ricca Chemical Company
RI70031 |
4 L | Black Poly Bottle |
Each for $1,858.44
|
|
||||
|
7003-5
|
Ricca Chemical Company
70035 |
20 L | Cubitainer |
Each for $7,489.64
|
|
||||
Chemical Identifiers
| 7761-88-8 | |
| 169.87 | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| 24470 | |
| silver(1+) nitrate |
| AgNO3 | |
| MFCD00003414 | |
| silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
| CHEBI:32130 | |
| [Ag+].[O-][N+]([O-])=O |
Specifications
| 7761-88-8,7732-18-5 | |
| 99.0,2.89 | |
| MFCD00003414 | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| silver(1+) nitrate | |
| 1 mL = 10 mg NaCl | |
| CHEBI:32130 | |
| 0.171N | |
| ∼6 | |
| Silver Nitrate |
| 95.0,2.77 | |
| AgNO3 | |
| silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
| [Ag+].[O-][N+]([O-])=O | |
| 169.87 | |
| 24470 | |
| Laboratory | |
| Amber Poly Bottle | |
| 500 mL |