missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate, 0.171 N, 1 mL = 10 mg NaCl, Ricca Chemical

For Chloride Titrations

Supplier:  Ricca Chemical Company RI70031

 View more versions of this product

Catalog No. 7003-1


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Silver Nitrate
95.0,2.77
AgNO3
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
[Ag+].[O-][N+]([O-])=O
169.87
24470
Laboratory
Black Poly Bottle
4 L
7761-88-8,7732-18-5
99.0,2.89
MFCD00003414
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver(1+) nitrate
1 mL = 10 mg NaCl
CHEBI:32130
0.171N
∼6

Chemical Identifiers

7761-88-8
169.87
SQGYOTSLMSWVJD-UHFFFAOYSA-N
24470
silver(1+) nitrate
AgNO3
MFCD00003414
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
CHEBI:32130
[Ag+].[O-][N+]([O-])=O
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions