missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Nitrate, 0.171 N, 1 mL = 10 mg NaCl, Ricca Chemical
For Chloride Titrations
Supplier: Ricca Chemical Company 700316
Specifications
| Silver Nitrate | |
| 95.0,2.77 | |
| AgNO3 | |
| silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
| [Ag+].[O-][N+]([O-])=O | |
| 169.87 | |
| 24470 | |
| Laboratory | |
| Amber Poly Bottle | |
| 500 mL |
| 7761-88-8,7732-18-5 | |
| 99.0,2.89 | |
| MFCD00003414 | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| silver(1+) nitrate | |
| 1 mL = 10 mg NaCl | |
| CHEBI:32130 | |
| 0.171N | |
| ∼6 |
Chemical Identifiers
| 7761-88-8 | |
| 169.87 | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| 24470 | |
| silver(1+) nitrate |
| AgNO3 | |
| MFCD00003414 | |
| silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
| CHEBI:32130 | |
| [Ag+].[O-][N+]([O-])=O |