missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate, 0.171 N, 1 mL = 10 mg NaCl, Ricca Chemical

Catalog No. 70035
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Amber Poly Bottle
Black Poly Bottle
Cubitainer
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
70035 20 L Cubitainer
700316 500 mL Amber Poly Bottle
700332 1 L Amber Poly Bottle
70031 4 L Black Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 70035 Supplier Ricca Chemical Company Supplier No. 70035
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Chloride Titrations

Chemical Identifiers

CAS 7761-88-8 , 7732-18-5
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

CAS Min % 95.0, 2.77
CAS Max % 99.0, 2.89
Equivalence 1 mL = 10 mg NaCl
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.171N
Packaging Cubitainer
pH ∼6
Quantity 20 L
Chemical Name or Material Silver Nitrate