missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Nitrate, 0.0141 N (0.0141 M), 1 mL = 0.5 mg Cl-, Ricca Chemical
Specifications
Specifications
| Color | Colorless |
| CAS Min % | 98.0, 0.23 |
| CAS Max % | 100.0, 0.24 |
| Equivalence | 1 mL = 0.5 mg Cl |
| Grade | Laboratory |
| Packaging | Black Poly Bottle |
| Methods | APHA for Chloride |
| pH | ∼6 |
| Quantity | 4 L |
| Physical Form | Liquid |
| Show More |