missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Nitrate, 0.0141 N (0.0141 M), 1 mL = 0.5 mg Cl-, Ricca Chemical
Supplier: Ricca Chemical Company 686025
Specifications
| Silver Nitrate | |
| 7761-88-8,7732-18-5 | |
| 100.0,0.24 | |
| MFCD00003414 | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| silver(1+) nitrate | |
| 1 mL = 0.5 mg Cl | |
| CHEBI:32130 | |
| Cubitainer | |
| ∼6 | |
| Liquid |
| Colorless | |
| 98.0,0.23 | |
| AgNO3 | |
| silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
| [Ag+].[O-][N+]([O-])=O | |
| 169.87 | |
| 24470 | |
| Laboratory | |
| APHA for Chloride | |
| 10 L | |
| 0.0141N,0.0141M |
Chemical Identifiers
| 7761-88-8 | |
| 169.87 | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| 24470 | |
| silver(1+) nitrate |
| AgNO3 | |
| MFCD00003414 | |
| silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
| CHEBI:32130 | |
| [Ag+].[O-][N+]([O-])=O |