missing translation for 'onlineSavingsMsg'
Learn More

Silver ICP Standard, 1000 ppm Ag in 3% HNO3, Ricca Chemical

Supplier:  Ricca Chemical Company PAG1KN500

 View more versions of this product

Catalog No. PAG1KN-500


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Silver ICP Standard
7761-88-8 , 7697-37-2 , 7732-18-5
94.0,4.22,0.15
1 mL = 1 mg Ag
AgNO3
Colorless
Approximately 0°C
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver(1+) nitrate
500 mL
CHEBI:32130
Amber Poly Bottle
1,000ppm Ag
97.0,4.39,0.15
Liquid
MFCD00003414
< 2
Standard
[Ag+].[O-][N+]([O-])=O
169.87
24470

Chemical Identifiers

7761-88-8
169.87
SQGYOTSLMSWVJD-UHFFFAOYSA-N
CHEBI:32130
[Ag+].[O-][N+]([O-])=O
AgNO3
MFCD00003414
24470
silver(1+) nitrate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions