missing translation for 'onlineSavingsMsg'
Learn More

Silver ICP Standard, 1000 ppm Ag in 3% HNO3, Ricca Chemical

Catalog No. p-3252259
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Amber Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PAG1KN100 100 mL Amber Poly Bottle
PAG1KN500 500 mL Amber Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PAG1KN100 Supplier Ricca Chemical Company Supplier No. PAG1KN100
Only null left

Chemical Identifiers

CAS 7761-88-8 , 7697-37-2 , 7732-18-5
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Packaging Amber Poly Bottle
Concentration or Composition Notes 1,000ppm Ag
CAS Min % 94.0, 4.22, 0.15
CAS Max % 97.0, 4.39, 0.15
Equivalence 1 mL = 1 mg Ag
Physical Form Liquid
Color Colorless
pH < 2
Melting Point Approximately 0°C
Type Standard
Quantity 100 mL
Chemical Name or Material Silver ICP Standard
Show More Show Less