missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silitech, 63-200μm, 60Å, MP Biomedicals™
For Chromatography; Chemical Synthesis
Supplier: MP Biomedicals Inc 0202066
Spécifications
| Silitech | |
| 7631-86-9 | |
| 25 kg | |
| MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
| VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
| silanedione | |
| 24261 | |
| 60.08 |
| White | |
| Powder | |
| O2Si | |
| silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
| O=[Si]=O | |
| 60.08 | |
| CHEBI:30563 | |
| 2g/cm3 at 20°C |
Identifiants chimiques
| 7631-86-9 | |
| 60.08 | |
| VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
| 24261 | |
| silanedione |
| O2Si | |
| MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
| silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
| CHEBI:30563 | |
| O=[Si]=O |
Sécurité et manipulation
Recommended Storage : Store at Room Temperature.