missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Silica gel, for chromatography, 0.035-0.070 mm, 60 A

Numéro de catalogue. AC240360300
Change view
Click to view available options
Quantity:
1 kg
5 kg
30 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AC240360300 30 kg
AC240360050 5 kg
AC240360010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AC240360300 Fournisseur Thermo Scientific Chemicals Code fournisseur 240360300
Il en reste null

For column chromatography. | CAS: 7631-86-9

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For column chromatography

Identifiants chimiques

CAS 7631-86-9

Spécifications

Pore Size 60 Å
Quantity 30 kg
Type Silica Gel
Material Silica Gel
For Use With (Application) Column Chromatography
Molecular Formula O2Si
Solubility Information Solubility aq. soln.: insoluble.
Formula Weight 60.08
Chemical Name or Material Silica gel

RUO – Research Use Only