| Numéro d'enregistrement |
P40189 |
| Plage de l'essai |
4-6000 pg/mL |
| Sensibilité d’essai |
4 pg/mL |
| Plage d’étalonnage |
4 pg/mLto 6000 pg/mL |
| Contenu et stockage |
Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability. |
| Essai |
Solid-phase sandwich ELISA |
| Conjugué |
HRP |
| Format de la trousse d’immunodosage |
Sandwich ELISA Kit |
| Type de produit |
ELISA |
| Famille de protéines |
Growth Factors & Receptors |
| Type d’échantillon |
Plasma, Serum, Supernatant |
| Spécificité |
This ELISA kit shows no cross-reactivity with any of the cytokines tested: Human Angiogenin, BDNF, BLC, ENA-78, FGF-4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-7, IL-8, IL-9, IL-10, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF, GM-CSF, IFN-gamma, Leptin, MCP-1, MCP-2, MCP-3, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF. |
| Suffisant pour |
96 Assays |
| Espèces cibles |
Human |
| À utiliser avec (application) |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis |
| À utiliser avec (équipement) |
Absorbance Microplate Reader |
| Alias du gène |
5133400A03Rik; AA389424; Ac1055; BB405851; CD130; CD130 antigen; CDW130; D13Ertd699e; EGK_16499; Gp130; gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form; gp130, oncostatin M receptor; GP130-RAPS; IL-6; IL-6 receptor subunit beta; IL-6R subunit beta; IL-6RB; IL-6R-beta; IL6R-beta; IL6ST; interleukin 6 signal transducer; interleukin 6 signal transducer (gp130, oncostatin M receptor); interleukin 6 signal transducer receptor; interleukin receptor beta chain; interleukin-6 receptor subunit beta; Interleukin-6 signal transducer; interleukin-6 signal transducer receptor; membrane glycoprotein; membrane glycoprotein 130; membrane glycoprotein gp130; oncostatin-M receptor subunit alpha5133400A03Rik; AA389424; Ac1055; BB405851; CD130; CD130 antigen; CDW130; D13Ertd699e; EGK_16499; Gp130; gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form; gp130, oncostatin M receptor; GP130-RAPS; IL-6; IL-6 receptor subunit beta; IL-6R subunit beta; IL-6RB; IL-6R-beta; IL6R-beta; IL6ST; interleukin 6 signal transducer; interleukin 6 signal transducer (gp130, oncostatin M receptor); interleukin 6 signal transducer receptor; interleukin receptor beta chain; interleukin-6 receptor subunit beta; Interleukin-6 signal transducer; interleukin-6 signal transducer receptor; membrane glycoprotein; membrane glycoprotein 130; membrane glycoprotein gp130; oncostatin-M receptor subunit alpha; CD130; CDW130; GP130; IL-6RB |
| ID du gène (Entrez) |
3572 |
| Symbole de gène |
IL6ST |
| CV intra-essai |
<12%, <10% |
| Contenus des kits |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers |
| Étiquette ou colorant |
Biotin |
| Quantité |
96 Tests |
| Statut réglementaire |
RUO |
| Disciplines de recherche |
Inflammation |
| Volume d’échantillon |
Plasma 1 μL, Serum 1 μL, Supernatant 100 μL |
| Conditions de stockage |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C |
| Synonyme |
CD130, CDW130, GP130, IL-6 RB |
| Cible |
GP130 |
| Durée du test |
1 hr 20 min |
| Durée totale de l'essai |
4 hr 45 min |
|
Afficher plus de résultats
Afficher moins de résultats
|
|