missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Shell Vials with Plugs

Vials are manufactured from chemically resistant, low extractable borosilicate glass

Supplier:  MilliporeSigma™ Supelco™ 24727

 Afficher plus de versions de ce produit

Numéro de catalogue. 111010947


Il en reste null

Description

Description

  • Have a uniform wall thickness and annealed top.
  • Natural color virgin polyethylene push-in caps are included in each kit.
  • These products meet or exceed original equipment manufacturer's requirements
Spécifications

Identifiants chimiques

8 mm
Clear Glass
0.75 mL
35 mm

Spécifications

8 mm
0.75 mL
200
Clear Glass
Displacement Plug
35 mm
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions