missing translation for 'onlineSavingsMsg'
Learn More

Cytiva SFX-Insect™ Cell Culture Media

Catalog No. SH30278LS
Change view
Click to view available options
Quantity:
1000 mL
6 x 1000 mL
With Additives:
Glutamine, Poloxamer 188
Glutamine, Sodium Bicarbonate, Poloxamer 188
Without Additives:
No Protein, No Phenol Red, No HEPES
Form:
Liquid
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity With Additives Without Additives Form
SH30278LS 6 x 1000 mL Glutamine, Sodium Bicarbonate, Poloxamer 188 No Protein, No Phenol Red, No HEPES Liquid
SH3027802 1000 mL Glutamine, Poloxamer 188 No Protein, No Phenol Red, No HEPES Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SH30278LS Supplier Cytiva Supplier No. SH30278.LS
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed to support the growth of multiple insect cell lines and production of a variety of recombinant proteins. HyClone™ SFX Insect™ Cell Culture Media is a protein-free media that support cell culture growth.

  • Protein free media

Specifications

Form Liquid
Product Type Insect Cell Medium
Sterilization Method 0.2 μm Sterile-filtered
With Additives Glutamine, Sodium Bicarbonate, Poloxamer 188
Without Additives No Protein, No Phenol Red, No HEPES
Quantity 6 x 1000 mL
Certifications/Compliance cGMP
Content And Storage 2° to 8°C, away from light