Learn More
Excelta™ Self-Opening Scissors
With simple squeeze action for precision cutting applications
Supplier:  Excelta™ 346B
 Afficher plus de versions de ce produit
Numéro de catalogue. 17467492
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Self-opening and closing system for cutting very thin soft wires
  • With stainless steel blade hardened to Rockwell hardness of C-50
  • Cut length: 0.63 in. (1.6cm) or 0.375 in. (9.5cm)
  • Blade length: 0.5 in. (1.3m)
Spécifications
Spécifications
Straight
3.5 in.
8.89 cm
Wire cutting
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS