Learn More
BD Veritor™ SARS-CoV-2 Control Swabs
10 SARS-CoV-2 positive and 10 SARS-CoV-2 negative quality control swabs
Supplier:  BD Veritor™ 256087
Numéro de catalogue. B256087
Il en reste null
Description
Description
  • To be used with BD Veritor™ System
  • For Rapid Detection of SARS-CoV-2 (256082)
Spécifications
Spécifications
SARS-CoV-2
BD Veritor System for Rapid Detection of SARS-CoV-2 (256082)
Control Swab
10 Paired Positive/Negative Control Swabs
Nasal Swab
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS
Documents
Documents
Promotions
Promotions
*The BD Veritor™ Plus System for Rapid Detection of SARS-CoV-2 is intended for the qualitative detection of SARS-CoV-2 nucleocapsid antigens in direct anterior nasal swabs from individuals who are either suspected of COVID-19 by their health care provider within the first five days of the onset of symptoms, or from individuals without symptoms or other epidemiological reasons to suspect COVID-19 when tested twice over two or three days with at least 24 hours and no more than 48 hours between tests.
  • This product has not been FDA cleared or approved; but has been authorized by FDA under an EUA for use by authorized laboratories.
  • This product has been authorized only for the detection of proteins from SARS-CoV-2, not for any other viruses or pathogens.
  • This product is only authorized for the duration of the declaration that circumstances exist justifying the authorization of emergency use of in vitro diagnostics for detection and/or diagnosis of COVID-19 under Section 564(b)(1) of the Federal Food, Drug and Cosmetic Act, 21 U.S.C. § 360bbb-3(b)(1), unless the declaration is terminated or authorization is revoked sooner.