Learn More
Thermo Scientific Chemicals Sarcosine, 98%
CAS: 107-97-1 | C3H7NO2 | 89.09 g/mol
Supplier: Thermo Scientific Chemicals A145940E
Description
A type 1 glycine transporter inhibitorSarcosine is used in the production of biodegradable surfactants and toothpastes. It is used as an intermediate in the synthesis of antienzyme agents. It is found to be a marker for prostate cancer bioagression. It acts as a biochemical reagent, dye stabilizer, daily chemical, amino acid surfactants, health drugs fatigue recovery agent and so on. It is involved in the preparation of N-methyl-glycine ethyl ester by reacting with ethanol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA type 1 glycine transporter inhibitorSarcosine is used in the production of biodegradable surfactants and toothpastes. It is used as an intermediate in the synthesis of antienzyme agents. It is found to be a marker for prostate cancer bioagression. It acts as a biochemical reagent, dye stabilizer, daily chemical, amino acid surfactants, health drugs fatigue recovery agent and so on. It is involved in the preparation of N-methyl-glycine ethyl ester by reacting with ethanol.
Solubility
Soluble in water, methanol. Slightly soluble in ether.
Notes
Incompatible with strong oxidizing agents.
Spécifications
| Sarcosine | |
| ∼208°C (decomposition) | |
| Odorless | |
| MFCD00004279 | |
| 1699442 | |
| 14,8373 | |
| Soluble in water,methanol. Slightly soluble in ether. | |
| CNCC(O)=O | |
| 89.09 | |
| CHEBI:15611 | |
| 98% |
| 107-97-1 | |
| 6.1 | |
| C3H7NO2 | |
| 2500 g | |
| Hygroscopic | |
| sarcosine, n-methylglycine, sarcosinic acid, methylglycine, glycine, n-methyl, methylaminoacetic acid, n-methylaminoacetic acid, sarcosin, 2-methylamino acetic acid, methylamino acetic acid | |
| FSYKKLYZXJSNPZ-UHFFFAOYSA-N | |
| 2-(methylamino)acetic acid | |
| 1088 | |
| 89.09 |
Identifiants chimiques
| 107-97-1 | |
| 89.09 | |
| FSYKKLYZXJSNPZ-UHFFFAOYSA-N | |
| 1088 | |
| 2-(methylamino)acetic acid |
| C3H7NO2 | |
| MFCD00004279 | |
| sarcosine, n-methylglycine, sarcosinic acid, methylglycine, glycine, n-methyl, methylaminoacetic acid, n-methylaminoacetic acid, sarcosin, 2-methylamino acetic acid, methylamino acetic acid | |
| CHEBI:15611 | |
| CNCC(O)=O |
Sécurité et manipulation
missing translation for 'einecsNumber' : 203-538-6
missing translation for 'rtecsNumber' : VQ2897000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only