Learn More
Chemglass Life Sciences 8mL (2 Dram) Vial, 17mm OD x 60mm Height, 15-425 Thread Size DFS Item
Supplier:  Chemglass Inc CG490002
8mL (2 Dram) Vial, 17mm OD x 60mm Height, 15-425 Thread Size. Sample vials packaged in convenient LAB-PACS of 144. Made from Type 1 Borosilicate glass. Full height partitions minimize breakage. Solid PTFE Lined Caps.
Numéro de catalogue. CG490002
Il en reste null
Découvrez plus d'offres spéciales