missing translation for 'onlineSavingsMsg'
Learn More

Samarium(III) 2,4-pentanedionate hydrate, 99.9% (REO)

CAS: 64438-53-5 | C15H21O6Sm | 447.69 g/mol

$69.56 - $190.66

Identifiants chimiques

CAS 64438-53-5
Molecular Formula C15H21O6Sm
Molecular Weight (g/mol) 447.69
MDL Number MFCD00792904
InChI Key JTZJMDRBZZVNSP-UHFFFAOYSA-N
Synonym samarium 2,4-pentanedionate, samarium iii acetylacetonate hydrate, samarium iii 2,4-pentanedionate hydrate, samarium iii acetylacetonate hydrate trace metals basis, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy samario oxy pent-3-en-2-one hydrate
PubChem CID 16213768
IUPAC Name (Z)-4-hydroxypent-3-en-2-one;samarium;hydrate
SMILES [Sm+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AA4066506
Afficher les documents
Thermo Scientific Chemicals
04066506
5 g
chaque for $69.56
Il en reste null
 
AA4066514
Afficher les documents
Thermo Scientific Chemicals
04066514
25 g
chaque for $190.66
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

64438-53-5
447.69
JTZJMDRBZZVNSP-UHFFFAOYSA-N
16213768
[Sm+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
C15H21O6Sm
MFCD00792904
samarium 2,4-pentanedionate, samarium iii acetylacetonate hydrate, samarium iii 2,4-pentanedionate hydrate, samarium iii acetylacetonate hydrate trace metals basis, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy samario oxy pent-3-en-2-one hydrate
(Z)-4-hydroxypent-3-en-2-one;samarium;hydrate

Spécifications

64438-53-5
5 g
MFCD00792904
JTZJMDRBZZVNSP-UHFFFAOYSA-N
(Z)-4-hydroxypent-3-en-2-one;samarium;hydrate
16213768
99.9%
(REO)
Powder
C15H21O6Sm
samarium 2,4-pentanedionate, samarium iii acetylacetonate hydrate, samarium iii 2,4-pentanedionate hydrate, samarium iii acetylacetonate hydrate trace metals basis, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy samario oxy pent-3-en-2-one hydrate
[Sm+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
447.69
447.68 (Anhydrous)
Samarium(III) 2,4-pentanedionate hydrate
Videos
Sécurité et manipulation

Sécurité et manipulation

missing translation for 'einecsNumber' : 238-632-6

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only