missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-Trichostatic Acid 98.0+%, TCI America™
Supplier: TCI America T3402200MG
Spécifications
| (S)-Trichostatic Acid | |
| 91°C | |
| C17H21NO3 | |
| (S,2E,4E)-7-[4-(Dimethylamino)phenyl]-4,6-dimethyl-7-oxo-2,4-heptadienoic Acid | |
| CC(C=C(C)C=CC(=O)O)C(=O)C1=CC=C(C=C1)N(C)C | |
| 287.359 | |
| CHEBI:39159 | |
| ≥98.0% (HPLC) |
| 68690-19-7 | |
| White-Yellow | |
| 200 mg | |
| VKEITMNFEJHFCX-LEJRBOCMSA-N | |
| (2E,4E,6S)-7-[4-(dimethylamino)phenyl]-4,6-dimethyl-7-oxohepta-2,4-dienoic acid | |
| 14100757 | |
| 287.36 | |
| Crystalline Powder |
Identifiants chimiques
| 68690-19-7 | |
| 287.359 | |
| (S,2E,4E)-7-[4-(Dimethylamino)phenyl]-4,6-dimethyl-7-oxo-2,4-heptadienoic Acid | |
| CHEBI:39159 | |
| CC(C=C(C)C=CC(=O)O)C(=O)C1=CC=C(C=C1)N(C)C |
| C17H21NO3 | |
| VKEITMNFEJHFCX-LEJRBOCMSA-N | |
| 14100757 | |
| (2E,4E,6S)-7-[4-(dimethylamino)phenyl]-4,6-dimethyl-7-oxohepta-2,4-dienoic acid |
Sécurité et manipulation
missing translation for 'tsca' : No
Recommended Storage : Refrigerator