missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rubidium fluoride, Puratronic™, 99.975% (metals basis), Thermo Scientific Chemicals
Description
As raw materials for preparation of rubidium metal and various rubidium salts, for the manufacturing of catalyst and for the manufacturing of high energy density micro cells and crystal scintillation counters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Spécifications
| Melting Point | 795°C |
| Color | White |
| Physical Form | Crystalline |
| Quantity | 10 g |
| Linear Formula | RbF |
| UN Number | UN3288 |
| Solubility Information | Soluble in water. Insoluble in ethanol. |
| Formula Weight | 104.47 |
| Percent Purity | 99.975% |
| Grade | Puratronic |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | H302+H312+H332-H315-H319-H335 |
| Hazard Statement | GHS H Statement H301-H311-H331-H315-H319-H335 Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S. |
| EINECSNumber | 236-603-2 |
| RTECSNumber | VL8740000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only