Learn More
Rock Formation Charts
$34.21 - $131.09
Produits 5
Numéro de catalogue Numéro du manufacturier. Product Type Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Product Type Prix Quantité  
S45002
Afficher les documents
Geoscience Industries
S45002
Sedimentary Rocks Chart
chaque for $131.09
Il en reste null
S45002A
Afficher les documents
Geoscience Industries
S45002A
Sedimentary Rock Reference Sheet
paquet de 15 chaque for $34.21
Il en reste null
S45004A
Afficher les documents
Geoscience Industries
S45004A
Metamorphic Rock Reference Sheet
paquet de 15 chaque for $34.21
Il en reste null
S45004
Afficher les documents
Geoscience Industries
S45004
Metamorphic Rocks Chart
chaque for $131.09
Il en reste null
S45018A
Afficher les documents
Geoscience Industries
S45018A
Igneous Rock Chart
paquet de 15 chaque for $34.21
Il en reste null
Description
Description
This coordinated series of display charts describes the classification, identification, and formation of the three major rock types. each chart also includes representative specimens of the appropriate rock group. For each of the charts, sets of 15 notebook-sized student reference sheets are also available.
Igneous Rocks Chart
A systematic arrangement of igneous rocks according to texture, composition, and origin. Includes 33 rock specimens.
Metamorphic Rocks Chart
The intricate processes by which a parent rock or mineral is changed (metamorphosed) to a new form are graphically illustrated. Includes 26 rock and mineral specimens.
Sedimentary Rocks Chart
The three main categories of sedimentary rocks (clastic, chemical, and organic) are clearly demonstrated with emphasis on the importance of origin and particle size. Includes 21 rock specimens.
Order Info
Measuring 60 x 39cm and mounted in wood frames.
Videos
FDS
missing translation for 'documents'
missing translation for 'documents'