Learn More
Epredia™ Resolve™ Immersion Oil
Eliminate toxicity and disposal problems with the Epredia™ Resolve™ Immersion Oil.
$33.71 - $1,071.89
| Catalog Number | Mfr. No. | Viscosity | Quantity | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Viscosity | Quantity | Price | Quantity & Availability | ||||
|
23244267
|
Epredia™
M5004 |
High | 16 oz. |
Each for $139.32
Case of 12 Each for $1,671.84
|
|
||||
|
1236519
|
Epredia™
M5000 |
Low | 16 oz. |
Each for $143.18
Case of 12 Each for $1,718.16
|
|
||||
|
1236520
|
Epredia™
M2000 |
Low | 0.5 oz. |
Each for $34.50
Case of 12 Each for $414.00
|
|
||||
|
23244265
|
Epredia™
M4004 |
High | 4 oz. |
Each for $53.13
Case of 12 Each for $637.56
|
|
||||
|
23244263
|
Epredia™
M3004 |
High | 1 oz. |
Each for $43.93
Case of 12 Each for $527.16
|
|
||||
|
23244264
|
Epredia™
M4000 |
Low | 4 oz. |
Case of 12 Each for $1,071.89
|
|
||||
|
23244262
|
Epredia™
M3000 |
Low | 1 oz. |
Each for $44.61
Case of 12 Each for $535.32
|
|
||||
|
123651
|
Epredia™
M2004 |
High | 0.5 oz. |
Each for $33.71
Case of 12 Each for $404.52
|
|
||||
Description
- Manufactured using a proprietary process that removes interfering substances and will not damage valuable optics or irreplaceable slide specimens
- Packaged conveniently in four sizes
Product Details:
- Available in both low and high viscosity
- Non-drying and non-hardening
- Refractive index of 1.5150
Specifications
| 1.515 |