missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Resolve™ Immersion Oil
Description
- Manufactured using a proprietary process that removes interfering substances and will not damage valuable optics or irreplaceable slide specimens
- Packaged conveniently in four sizes
Product Details:
- Available in both low and high viscosity
- Non-drying and non-hardening
- Refractive index of 1.5150
Specifications
Specifications
| Type | Immersion Oil |
| Viscosity | High |
| Quantity | 0.5 oz. |
| Refractive Index | 1.515 |