missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Resolve™ Immersion Oil

Catalog No. 1236520
Change view
Click to view available options
Viscosity:
High
Low
Quantity:
0.5 oz.
1 oz.
4 oz.
16 oz.
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Viscosity Quantity
1236520 Low 0.5 oz.
23244267 High 16 oz.
1236519 Low 16 oz.
23244265 High 4 oz.
23244263 High 1 oz.
23244264 Low 4 oz.
23244262 Low 1 oz.
123651 High 0.5 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 1236520 Supplier Epredia™ Supplier No. M2000
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Eliminate toxicity and disposal problems with the Epredia™ Resolve™ Immersion Oil.

  • Manufactured using a proprietary process that removes interfering substances and will not damage valuable optics or irreplaceable slide specimens
  • Packaged conveniently in four sizes

Product Details:

  • Available in both low and high viscosity
  • Non-drying and non-hardening
  • Refractive index of 1.5150

Specifications

Viscosity Low
Quantity 0.5 oz.
Refractive Index 1.515