Learn More
Epredia™ Resolve™ Immersion Oil
Eliminate toxicity and disposal problems with the Epredia™ Resolve™ Immersion Oil.
Supplier:  Epredia™ M4000
 View more versions of this product
Catalog No. 23244264
Only null left
Description
Description
  • Manufactured using a proprietary process that removes interfering substances and will not damage valuable optics or irreplaceable slide specimens
  • Packaged conveniently in four sizes
Product Details:
  • Available in both low and high viscosity
  • Non-drying and non-hardening
  • Refractive index of 1.5150
Specifications
Chemical Identifiers
1.515
Specifications
Immersion Oil
Low
4 oz.
1.515
Product Suggestions
Product Suggestions