Learn More
Epredia™ Resolve™ Immersion Oil
Eliminate toxicity and disposal problems with the Epredia™ Resolve™ Immersion Oil.
Supplier: Epredia™ M5000
Description
- Manufactured using a proprietary process that removes interfering substances and will not damage valuable optics or irreplaceable slide specimens
- Packaged conveniently in four sizes
Product Details:
- Available in both low and high viscosity
- Non-drying and non-hardening
- Refractive index of 1.5150
Chemical Identifiers
| 1.515 |
Specifications
| Immersion Oil | |
| Low | |
| 16 oz. | |
| 1.515 |