missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Resolve™ Immersion Oil

Catalog No. 23244265
Change view
Click to view available options
Viscosity:
High
Low
Quantity:
0.5 oz.
1 oz.
4 oz.
16 oz.
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Viscosity Quantity
23244265 High 4 oz.
23244267 High 16 oz.
1236519 Low 16 oz.
1236520 Low 0.5 oz.
23244263 High 1 oz.
23244264 Low 4 oz.
23244262 Low 1 oz.
123651 High 0.5 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 23244265 Supplier Epredia™ Supplier No. M4004
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Eliminate toxicity and disposal problems with the Epredia™ Resolve™ Immersion Oil.

  • Manufactured using a proprietary process that removes interfering substances and will not damage valuable optics or irreplaceable slide specimens
  • Packaged conveniently in four sizes

Product Details:

  • Available in both low and high viscosity
  • Non-drying and non-hardening
  • Refractive index of 1.5150

Specifications

Type Immersion Oil
Viscosity High
Quantity 4 oz.
Refractive Index 1.515