missing translation for 'onlineSavingsMsg'
Learn More

Modified Davidson's Fixative II, Ricca Chemical

Catalog No. p-4842623
Change view
Click to view available options
Quantity:
500 mL
55 gal.
Packaging:
Natural Poly Bottle
Poly Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
518716 500 mL Natural Poly Bottle
518755 55 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 518716 Supplier Ricca Chemical Company Supplier No. 518716

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Physical Form Solution
Packaging Natural Poly Bottle
Color Blue
pH 2.41 to 2.54
Quantity 500 mL
Grade Laboratory