missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G-Biosciences Reverse Transcriptase (RT)
Description
- Reverse Transcriptase (RT) is a recombinant Moloney Murine Leukemia Virus (MMLV) reverse transcriptase with reduced RNase H activity.
- This RT is an RNA-directed DNA polymerase which lacks 3' - 5' exonuclease activity and can produce full-length cDNA from long RNA templates.
Spécifications
Spécifications
| Detection Method | qPCR |
| Form | Liquid |
| GC-Rich PCR Performance | Medium |
| PCR Method | qPCR, Multiplex qPCR, 1-step RT-qPCR, 2-step RT-qPCR, 1-step RT-PCR |
| Reaction Speed | Fast or Standard |
| Label or Dye | Biotin, FAM, SYBR Green, ROX, TAMRA |
| Product Type | Reverse Transcriptase |
| For Use With (Application) | Gene Expression, Microbial Detection, Pathogen Detection, Virus Detection, cDNA Synthesis |
| No. of Reactions | 50,000 |