Learn More
Restek™ Ferrule Removal Kit DFS Item
Remove fused silica adaptor ferrules with this kit. Each Restek™ Ferrule Removal Kit has two tools: One for removing 1/32 inch adaptor ferrules and one for removing 1/16 inch adaptor ferrules.
Supplier:  Restek™ RK20146
Numéro de catalogue. 06710513
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • The tapered tools in this kit have teeth designed to grip and remove fused silica adaptor ferrules that have become stuck in the fitting detail
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS