Learn More
Thermo Scientific™ RBS™ pF Detergent Concentrate
A phosphate-free, surfactant-based cleaning solution for use on many laboratory surfaces, labware utensils and glassware.
$167.72 - $348.97
Spécifications
| Form | Liquid |
|---|---|
| Product Line | RBS |
| Composition | 2% (v/v) Solution |
| Content And Storage | Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25°C. |
| Description | RBS-pF Detergent Concentrate |
| Numéro de catalogue | Numéro du manufacturier. | Volume (Metric) | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Volume (Metric) | Prix | Quantity & Availability | |||||
|
PI27959
|
Thermo Scientific™
27959 |
1 kg |
paquet for $167.72
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
PI27960
|
Thermo Scientific™
27960 |
5 kg |
paquet for $348.97
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
Thermo Scientific™ RBS™-pF Detergent Concentrate is a phosphate-free formulation of a surfactant-based cleaning agent for use on many laboratory surfaces and glassware.
Highlights:
- Ideal for removing radioactivity on contaminated bench surfaces
- Low-foaming
- Rinses easily without leaving residues
- Fully biodegradable
- Phosphate free
Applications of RBS pF Detergent Concentrate include the removal of:
- Coomassie blue dye from test tubes and quartz cuvettes
- Blood or serum residue from glass or plastic beakers and flasks
- Colored dyes from porcelain mortar and pestles
- HIV virus from glassware, tabletops and flooring
- Radioactive isotopes from benchtops and instrument surfaces
The cleaning detergent is an alkaline solution of anionic and nonionic surfactants. RBS™ pF Detergent is strong enough to remove biological residues from plastic and glass, yet it is fully biodegradable and does not contain phosphates that can harm the environment. Use a 2% (v/v) solution for routine washing applications or soaking in a warm 20% solution to free particularly stubborn dry residues.
Complete removal of all impurities and contaminants from glassware is a necessity for most laboratory applications. For many years, caustic alcohol and chromic acid solutions were used for many substances that could not be cleaned from glassware by ordinary detergents. These strongly acidic or strongly basic cleaning products reduce the lifespan of glassware while exposing the user to potential harm. By contrast, Thermo Scientific RBS 35™ and PCC-54 Products are mildly basic, biodegradable, surfactant concentrates containing anionic and nonionic detergents. The surfactants clean by penetrating between contaminating substances and the surfaces to be cleaned. Glass, plastics, quartz, porcelain and ferrous metal apparatus can be cleaned safely and effectively in a 2% solution (v/v) of the concentrates or a 0.2% solution (w/v) of the solids. RBS and PCC-54 Cleaners remove both organic and inorganic materials and metal ions from laboratory equipment. The detergents rinse away completely, leaving no residue.
Note:
We do not sell RBS Products in Europe; EU customers can purchase our PCC-54 Products instead.
Spécifications
| Liquid | |
| 2% (v/v) Solution | |
| RBS-pF Detergent Concentrate |
| RBS | |
| Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25°C. |
For Research Use Only. Not for use in diagnostic procedures.