missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RBS™ pF Detergent Concentrate

Catalog No. PI27959
Change view
Click to view available options
Volume (Metric):
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
PI27959 1 kg
PI27960 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI27959 Supplier Thermo Scientific™ Supplier No. 27959

A phosphate-free, surfactant-based cleaning solution for use on many laboratory surfaces, labware utensils and glassware.

Thermo Scientific™ RBS™-pF Detergent Concentrate is a phosphate-free formulation of a surfactant-based cleaning agent for use on many laboratory surfaces and glassware.

Highlights:

  • Ideal for removing radioactivity on contaminated bench surfaces
  • Low-foaming
  • Rinses easily without leaving residues
  • Fully biodegradable
  • Phosphate free

Applications of RBS pF Detergent Concentrate include the removal of:

  • Coomassie blue dye from test tubes and quartz cuvettes
  • Blood or serum residue from glass or plastic beakers and flasks
  • Colored dyes from porcelain mortar and pestles
  • HIV virus from glassware, tabletops and flooring
  • Radioactive isotopes from benchtops and instrument surfaces

The cleaning detergent is an alkaline solution of anionic and nonionic surfactants. RBS™ pF Detergent is strong enough to remove biological residues from plastic and glass, yet it is fully biodegradable and does not contain phosphates that can harm the environment. Use a 2% (v/v) solution for routine washing applications or soaking in a warm 20% solution to free particularly stubborn dry residues.

Complete removal of all impurities and contaminants from glassware is a necessity for most laboratory applications. For many years, caustic alcohol and chromic acid solutions were used for many substances that could not be cleaned from glassware by ordinary detergents. These strongly acidic or strongly basic cleaning products reduce the lifespan of glassware while exposing the user to potential harm. By contrast, Thermo Scientific RBS 35™ and PCC-54 Products are mildly basic, biodegradable, surfactant concentrates containing anionic and nonionic detergents. The surfactants clean by penetrating between contaminating substances and the surfaces to be cleaned. Glass, plastics, quartz, porcelain and ferrous metal apparatus can be cleaned safely and effectively in a 2% solution (v/v) of the concentrates or a 0.2% solution (w/v) of the solids. RBS and PCC-54 Cleaners remove both organic and inorganic materials and metal ions from laboratory equipment. The detergents rinse away completely, leaving no residue.

Note:

We do not sell RBS Products in Europe; EU customers can purchase our PCC-54 Products instead.

Specifications

Form Liquid
Product Line RBS
Composition 2% (v/v) Solution
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25°C.
Description RBS-pF Detergent Concentrate
Quantity 1 kg
Volume (Metric) 1 kg
Are RBS 35 Detergent Concentrate and PCC-54 Detergent Concentrate phosphate-free?

RBS-pF Detergent Concentrate (Cat. Nos. 27959 and 27960), and PCC-Pfree Detergent Concentrate (Cat. No. 72290) are all phosphate-free.

For Research Use Only. Not for use in diagnostic procedures.