Learn More
Pyrrole-2-carboxylic acid, 97%
CAS: 634-97-9 | C5H5NO2 | 111.1 g/mol
$219.53 - $219.53
Identifiants chimiques
| CAS | 634-97-9 |
|---|---|
| Molecular Formula | C5H5NO2 |
| Molecular Weight (g/mol) | 111.1 |
| MDL Number | MFCD00005219 |
| InChI Key | WRHZVMBBRYBTKZ-UHFFFAOYSA-N |
| Synonym | pyrrole-2-carboxylic acid, minaline, minalin, 2-pyrrolecarboxylic acid, pyrrole-2-carboxylicacid, pyrrole-2-carboxylate, 2-carboxypyrrole, 1h-pyrrolecarboxylic acid, 9ci, pyc |
| PubChem CID | 12473 |
| ChEBI | CHEBI:36751 |
| IUPAC Name | 1H-pyrrole-2-carboxylic acid |
| SMILES | C1=CNC(=C1)C(=O)O |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|
AC132070050
|
Thermo Scientific Chemicals
132070050 |
5 g | Glass bottle |
chaque for $219.53
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
| 634-97-9 | |
| 111.1 | |
| WRHZVMBBRYBTKZ-UHFFFAOYSA-N | |
| 12473 | |
| 1H-pyrrole-2-carboxylic acid |
| C5H5NO2 | |
| MFCD00005219 | |
| pyrrole-2-carboxylic acid, minaline, minalin, 2-pyrrolecarboxylic acid, pyrrole-2-carboxylicacid, pyrrole-2-carboxylate, 2-carboxypyrrole, 1h-pyrrolecarboxylic acid, 9ci, pyc | |
| CHEBI:36751 | |
| C1=CNC(=C1)C(=O)O |
Spécifications
| 634-97-9 | |
| Pink to White | |
| Authentic | |
| Glass bottle | |
| 1.4410 to 1.4450 | |
| 5 g | |
| pyrrole-2-carboxylic acid, minaline, minalin, 2-pyrrolecarboxylic acid, pyrrole-2-carboxylicacid, pyrrole-2-carboxylate, 2-carboxypyrrole, 1h-pyrrolecarboxylic acid, 9ci, pyc | |
| WRHZVMBBRYBTKZ-UHFFFAOYSA-N | |
| 1H-pyrrole-2-carboxylic acid | |
| 12473 | |
| 111.1 | |
| Powder |
| 207.0°C to 208.0°C | |
| 0.862g/mL | |
| 97% | |
| C5H5NO2 | |
| MFCD00005219 | |
| 22, 22 | |
| Solubility in water: soluble. Other solubilities: soluble in ethanol, diethyl ether | |
| C1=CNC(=C1)C(=O)O | |
| 111.1 | |
| CHEBI:36751 | |
| 97% | |
| Pyrrole-2-carboxylic acid |
Sécurité et manipulation
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 211-221-9
RUO – Research Use Only